accessibility menu, dialog, popup

UserName

CAS RN 205309-81-5

OOOHNH2OHO

CAS RN 205309-81-5

IUPAC Name: 2-amino-3-(benzyloxy)butanedioic acid
Synonyms: 2-amino-3-(benzyloxy)butanedioic acid
Molecular Weight (g/mol): 239.23
Molecular Formula: C11H13NO5
InChi Key: BYOBCYXURWDEDS-UHFFFAOYNA-N
SMILES: NC(C(OCC1=CC=CC=C1)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results