accessibility menu, dialog, popup

UserName

CAS RN 20567-38-8

H3COOOOH3CNOO(E)

CAS RN 20567-38-8

IUPAC Name: (2E)-3-(4,5-dimethoxy-2-nitrophenyl)prop-2-enoate
Synonyms: (2E)-3-(4,5-dimethoxy-2-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol): 252.2
Molecular Formula: C11H10NO6
InChi Key: BZIRMMAJZSOLEW-ONEGZZNKSA-M
SMILES: COC1=CC(\C=C\C([O-])=O)=C(C=C1OC)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results