Learn More
4,5-Dimethoxy-2-nitrocinnamic acid, 98%, Thermo Scientific™
Product Code. 11452498
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452498 25 g 25g
11442498 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11452498 Supplier Thermo Scientific Alfa Aesar Supplier No. A15824.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20567-38-8
Molecular Formula C11H11NO6
Molecular Weight (g/mol) 253.21
MDL Number MFCD00017017
InChI Key BZIRMMAJZSOLEW-ONEGZZNKSA-N
Synonym 3-4,5-dimethoxy-2-nitrophenyl acrylic acid, 3,4-dimethoxy-6-nitrocinnamic acid, 4,5-dimethoxy-2-nitrocinnamic acid, 2e-3-4,5-dimethoxy-2-nitrophenyl prop-2-enoic acid, 2-nitro-4,5-dimethoxycinnamic acid, 3-3,4-dimethoxy-6-nitrophenyl propenoic acid, e-3-4,5-dimethoxy-2-nitrophenyl prop-2-enoic acid
PubChem CID 735832
SMILES COC1=C(C=C(C(=C1)C=CC(=O)O)[N+](=O)[O-])OC
Specifications
Melting Point ∼293°C (decomposition)
Quantity 25 g
Formula Weight 253.21
Percent Purity 0.98
Chemical Name or Material 4,5-Dimethoxy-2-nitrocinnamic acid