CAS RN 207390-14-5
CAS RN 207390-14-5
IUPAC Name:
5-bromo-7,11-diazatricyclo[7.3.1.0²,⁷]trideca-2,4-dien-6-one
Synonyms:
3-bromocytisine
Molecular Weight (g/mol):
269.14
Molecular Formula:
C11H13BrN2O
InChi Key:
DWDCLEHDNICBMI-UHFFFAOYNA-N
SMILES:
BrC1=CC=C2C3CNCC(C3)CN2C1=O
1
–
1
of
1
results
1
Tocris Bioscience™ 3-Bromocytisine
CAS: 207390-14-5 Molecular Formula: C11H13BrN2O