missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 3-Bromocytisine
GREENER_CHOICE

Product Code. 18075412
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18075412

Brand: Tocris Bioscience™ 3549/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent agonist of α4β4, α4β2 and α7 nACh receptors

Potent agonist of α4β4, α4β2 and α7 nACh receptors (IC50 values are 0.28, 0.30 and 31.6 nM respectively). Displays different effects on high (HS) and low (LS) ACh sensitivity α4β2 nAChRs (EC50 values are 0.008 and 0.05 μM respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207390-14-5
Molecular Formula C11H13BrN2O

Specifications

Quantity 50 mg
Formula Weight 269.14
Purity >98%
Molecular Weight (g/mol) 269.142
Chemical Name or Material 3-Bromocytisine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.