accessibility menu, dialog, popup

UserName

CAS RN 2077-46-5

CH3ClClCl

CAS RN 2077-46-5

IUPAC Name: 1,2,4-trichloro-3-methylbenzene
Synonyms: 2,3,6-trichlorotoluene
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: UZYYBZNZSSNYSA-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC(Cl)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results