missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trichlorotoluene, TRC

Product Code. 30132410
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132410 1 g 1g
30006125 100 mg 100mg
30006523 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132410 Supplier Toronto Research Chemicals Supplier No. TRCT3532651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2077-46-5
Molecular Formula C7 H5 Cl3
Molecular Weight (g/mol) 195.47
Synonym Toluene, 2,3,6-trichloro- (7CI,8CI), 1,2,4-Trichloro-3-methylbenzene, 2,3,6-Trichlorotoluene
IUPAC Name 1,2,4-trichloro-3-methylbenzene
SMILES Cc1c(Cl)ccc(Cl)c1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5Cl3/c1-4-5(8)2-3-6(9)7(4)10/h2-3H,1H3
Formula Weight 193.9457
Chemical Name or Material 2,3,6-Trichlorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.