CAS RN 207801-29-4
IUPAC Name:
tris(4-hydroxypent-3-en-2-one) hydrate yttrium
Synonyms:
tris(4-hydroxypent-3-en-2-one) hydrate yttrium
Molecular Weight (g/mol):
407.27
Molecular Formula:
C15H26O7Y
InChi Key:
XTPGSXBDDAFYGV-UHFFFAOYSA-N
SMILES:
O.[Y].CC(O)=CC(C)=O.CC(O)=CC(C)=O.CC(O)=CC(C)=O
1
–
1
of
1
results
1
Yttrium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
CAS: 207801-29-4 Molecular Formula: C15H21O6Y Molecular Weight (g/mol): 386.23 MDL Number: MFCD00013509 InChI Key: CKIVBSKECCTIAG-UHFFFAOYSA-N Synonym: tris acetylacetonato yttrium n-hydrate,3e-4-bis 2e-4-oxopent-2-en-2-yl oxy yttrio oxy pent-3-en-2-one hydrate PubChem CID: 90471001 SMILES: [Y+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O