accessibility menu, dialog, popup

UserName

CAS RN 210688-56-5

H3CNNOHNCH3HClHCl

CAS RN 210688-56-5

IUPAC Name: 3-{2-[4-(3,4-dimethylphenyl)piperazin-1-yl]ethyl}-2,3-dihydro-1H-isoindol-1-one dihydrochloride
Synonyms: 3-{2-[4-(3,4-dimethylphenyl)piperazin-1-yl]ethyl}-2,3-dihydroisoindol-1-one dihydrochloride
Molecular Weight (g/mol): 422.39
Molecular Formula: C22H29Cl2N3O
InChi Key: RMNWLEGGYCVHFD-UHFFFAOYNA-N
SMILES: Cl.Cl.CC1=C(C)C=C(C=C1)N1CCN(CCC2NC(=O)C3=CC=CC=C23)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results