missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PD 168568 dihydrochloride
GREENER_CHOICE

Product Code. 18044732
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18044732

Brand: Tocris Bioscience™ 3529/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective D4 antagonist

Potent and selective dopamine D4 receptor antagonist (Ki values are 8.8 and 1842 nM at D4 and D2 receptors respectively). Reverses amphetamine-stimulated locomotion in vivo and is orally active.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210688-56-5
Molecular Formula C22H29Cl2N3O

Specifications

Quantity 50 mg
Formula Weight 422.39
Purity >98%
Molecular Weight (g/mol) 422.394
Chemical Name or Material PD 168568 dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.