accessibility menu, dialog, popup

UserName

CAS RN 21293-29-8

CH3(S)HOOH3CH3CCH3HOO(E)(Z)

CAS RN 21293-29-8

IUPAC Name: (2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienoic acid
Synonyms: abscisic acid (+)-abscisic acid abscisin II 2-cis,4-trans-abscisic acid
Molecular Weight (g/mol): 264.32
Molecular Formula: C15H20O4
InChi Key: JLIDBLDQVAYHNE-YKALOCIXSA-N
SMILES: C\C(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results