accessibility menu, dialog, popup

UserName

CAS RN 21306-21-8

ClClOClClClCl

CAS RN 21306-21-8

IUPAC Name: hexachlorocyclohexa-2,4-dien-1-one
Synonyms: hexachlorocyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 300.77
Molecular Formula: C6Cl6O
InChi Key: BBLJNWQYENOWPH-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(=O)C(Cl)(Cl)C(Cl)=C1Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results