CAS RN 21343-40-8
CAS RN 21343-40-8
IUPAC Name:
(1S,3Z)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5S)-6-hydroxy-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
Synonyms:
(1S,3Z)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5S)-6-hydroxy-5,6-dimethylhept-3-en-2-yl]-7a-methyl-hexahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
Molecular Weight (g/mol):
412.66
Molecular Formula:
C28H44O2
InChi Key:
KJKIIUAXZGLUND-ICCVIKJNSA-N
SMILES:
C[C@H](\C=C\[C@H](C)C(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)CCC1=C
1
–
4
of
4
results
1
25-Hydroxy Vitamin D2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
25-Hydroxy Vitamin D2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
25-Hydroxy Vitamin D2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Ercalcidiol, MedChemExpress
MedChemExpress Ercalcidiol is a metabolite of vitamin D2, is regarded as an indicator of vitamin D nutritional status.