CAS RN 2137047-43-7
CAS RN 2137047-43-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ VU 6005649
Positive allosteric modulators of mGlu7 and mGlu8 receptors. Also NK1 antagonist.
2
VU6005649, MedChemExpress
MedChemExpress VU6005649 is a CNS penetrant mGlu7/8 receptor agonist with EC50s of 0.65 μM and 2.6 μM for mGlu7 receptor and mGlu8 receptor, respectively.