CAS RN 2143949-52-2
CAS RN 2143949-52-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Keto-gestodene, TRC
CAS: 2143949-52-2 Molecular Formula: C21H24O3 Molecular Weight (g/mol): 324.41 Synonym: (8R,9S,10R,13S,14S,17R)-13-Ethyl-17-ethynyl-17-hydroxy-7,8,10,11,12,13,14,17-octahydro-1H-cyclopenta[a]phenanthrene-3,6(2H,9H)-dione IUPAC Name: (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-2,7,8,9,10,11,12,14-octahydro-1H-cyclopenta[a]phenanthrene-3,6-dione SMILES: CC[C@]12CC[C@H]3[C@@H](CC(=O)C4=CC(=O)CC[C@H]34)[C@@H]1C=C[C@@]2(O)C#C