missing translation for 'onlineSavingsMsg'
Learn More

6-Keto-gestodene, TRC

Product Code. 30407055
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407055 1 mg
30399024 5 mg
30399238 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407055 Supplier Toronto Research Chemicals Supplier No. TRCK1899701MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2143949-52-2
Molecular Formula C21H24O3
Molecular Weight (g/mol) 324.41
Synonym (8R,9S,10R,13S,14S,17R)-13-Ethyl-17-ethynyl-17-hydroxy-7,8,10,11,12,13,14,17-octahydro-1H-cyclopenta[a]phenanthrene-3,6(2H,9H)-dione
IUPAC Name (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-2,7,8,9,10,11,12,14-octahydro-1H-cyclopenta[a]phenanthrene-3,6-dione
SMILES CC[C@]12CC[C@H]3[C@@H](CC(=O)C4=CC(=O)CC[C@H]34)[C@@H]1C=C[C@@]2(O)C#C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H24O3/c1-3-20-9-7-15-14-6-5-13(22)11-17(14)19(23)12-16(15)18(20)8-10-21(20,24)4-2/h2,8,10-11,14-16,18,24H,3,5-7,9,12H2,1H3/t14-,15-,16-,18+,20+,21+/m1/s1
Formula Weight 324.1725
Chemical Name or Material 6-Keto-gestodene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.