accessibility menu, dialog, popup

UserName

CAS RN 218301-22-5

ONF

CAS RN 218301-22-5

IUPAC Name: 2-fluoro-5-formylbenzonitrile
Synonyms: 2-fluoro-5-formylbenzonitrile
Molecular Weight (g/mol): 149.12
Molecular Formula: C8H4FNO
InChi Key: MOFRJTLODZILCR-UHFFFAOYSA-N
SMILES: FC1=C(C=C(C=O)C=C1)C#N
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results