accessibility menu, dialog, popup

UserName

CAS RN 220227-37-2

HOFFF

CAS RN 220227-37-2

IUPAC Name: (3,4,5-trifluorophenyl)methanol
Synonyms: (3,4,5-trifluorophenyl)methanol
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: HRSFRSLKOPFWMZ-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=C(F)C(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results