missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzyl alcohol, 97%, Thermo Scientific™

Product Code. 11470478
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470478 5 g 5g
11460478 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470478 Supplier Thermo Scientific Alfa Aesar Supplier No. B24617.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,4,5-Trifluorobenzyl alcohol is used as chemical reagent.

Solubility
Soluble in water 100 g/L.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220227-37-2
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.11
MDL Number MFCD00083527
InChI Key HRSFRSLKOPFWMZ-UHFFFAOYSA-N
PubChem CID 2777040
IUPAC Name (3,4,5-trifluorophenyl)methanol
SMILES OCC1=CC(F)=C(F)C(F)=C1

Specifications

Density 1.385
Boiling Point 203°C to 204°C
Flash Point 98°C (208°F)
Refractive Index 1.4725
Quantity 5 g
Beilstein 8681069
Synonym 3,4,5-trifluorobenzyl alcohol, 3,4,5-trifluorophenyl methanol, 3,4,5-trifluorobenzenemethanol, benzenemethanol, 3,4,5-trifluoro, 3,4,5-trifluorophenyl methan-1-ol, 3,4,5-trifluoro benzenemethanol, acmc-20angf, 3,4,5-trifluorobenzylalcohol, 3,4,5-trifluoro-benzenemethanol, benzenemethanol,3,4,5-trifluoro
Solubility Information Soluble in water 100g/L.
Formula Weight 162.11
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzyl alcohol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.