accessibility menu, dialog, popup

UserName

CAS RN 220227-93-0

IUPAC Name: 1-[2-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[2-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: XTXSTESGCXKUIH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1OC(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results