CAS RN 22362-66-9
CAS RN 22362-66-9
IUPAC Name:
1-(3,5-dibromo-2-hydroxyphenyl)ethan-1-one
Synonyms:
1-(3,5-dibromo-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol):
293.94
Molecular Formula:
C8H6Br2O2
InChi Key:
RCKHUOIKVIOMGZ-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(Br)=CC(Br)=C1O
1
–
1
of
1
results
1
3',5'-Dibromo-2'-hydroxyacetophenone, 99%
CAS: 22362-66-9 Molecular Formula: C8H6Br2O2 Molecular Weight (g/mol): 293.942 MDL Number: MFCD00191992 InChI Key: RCKHUOIKVIOMGZ-UHFFFAOYSA-N Synonym: 3',5'-dibromo-2'-hydroxyacetophenone,1-3,5-dibromo-2-hydroxyphenyl ethanone,1-3,5-dibromo-2-hydroxyphenyl ethan-1-one,3´,5´-dibromo-2´-hydroxyacetophenone,1-acetyl-3,5-dibromo-2-hydroxybenzene,acmc-1cp7q,2-acetyl-4,6-dibromophenol,dibromooxochromenecarboxylicacid,3/',5/'-dibromo-2/'-hydroxyacetophenone,ethanone,1-3,5-dibromo-2-hydroxyphenyl PubChem CID: 271762 IUPAC Name: 1-(3,5-dibromo-2-hydroxyphenyl)ethanone SMILES: CC(=O)C1=CC(=CC(=C1O)Br)Br