accessibility menu, dialog, popup

UserName

CAS RN 2243-81-4

H2NO

CAS RN 2243-81-4

IUPAC Name: naphthalene-1-carboxamide
Synonyms: 1-naphthalenecarboxamide
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: RMHJJUOPOWPRBP-UHFFFAOYSA-N
SMILES: NC(=O)C1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)