accessibility menu, dialog, popup

UserName

NHNHH2N

CAS RN 22763-69-5

IUPAC Name: 4-[2-(pyrrolidin-1-ium-1-yl)ethyl]piperazine-1,4-diium
Synonyms: 4-[2-(pyrrolidin-1-ium-1-yl)ethyl]piperazine-1,4-diium
Molecular Weight (g/mol): 186.32
Molecular Formula: C10H24N3
InChi Key: UXHZIFKPLFIPSX-UHFFFAOYSA-Q
SMILES: C(C[NH+]1CC[NH2+]CC1)[NH+]1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results