CAS RN 23708-56-7
IUPAC Name:
undecan-6-ol
Synonyms:
6-undecanol
Molecular Weight (g/mol):
172.31
Molecular Formula:
C11H24O
InChi Key:
YBIXBBGRHOUVBB-UHFFFAOYSA-N
SMILES:
CCCCCC(O)CCCCC
1
–
3
of
3
results
1
6-Undecanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Undecanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Undecanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.