accessibility menu, dialog, popup

UserName

CAS RN 2388-10-5

LiH3CH3CO

CAS RN 2388-10-5

IUPAC Name: lithium(1+) propan-2-olate
Synonyms: lithium(1+) propan-2-olate
Molecular Weight (g/mol): 66.03
Molecular Formula: C3H7LiO
InChi Key: HAUKUGBTJXWQMF-UHFFFAOYSA-N
SMILES: [Li+].CC(C)[O-]
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)