accessibility menu, dialog, popup

UserName

CAS RN 240139-82-6

BFFFOCH3HOOH

CAS RN 240139-82-6

IUPAC Name: [2-methoxy-5-(trifluoromethyl)phenyl]boronic acid
Synonyms: 2-methoxy-5-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 219.95
Molecular Formula: C8H8BF3O3
InChi Key: PXDQUMRSSQYJSX-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)C(F)(F)F)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results