CAS RN 240401-22-3
CAS RN 240401-22-3
IUPAC Name:
piperidine-4-carbonitrile hydrochloride
Synonyms:
piperidine-4-carbonitrile hydrochloride
Molecular Weight (g/mol):
146.62
Molecular Formula:
C6H11ClN2
InChi Key:
PKMPMUHWHIIFBJ-UHFFFAOYSA-N
SMILES:
Cl.N#CC1CCNCC1
1
–
1
of
1
results
1
4-Cyanopiperidine hydrochloride, 97%, Thermo Scientific™
CAS: 240401-22-3 Molecular Formula: C6H10N2·ClH Molecular Weight (g/mol): 146.62 InChI Key: PKMPMUHWHIIFBJ-UHFFFAOYSA-N Synonym: piperidine-4-carbonitrile hydrochloride,4-cyanopiperidine hcl,4-cyanopiperidine hydrochloride,4-piperidinecarbonitrile, monohydrochloride,pubchem23464,4-cyanopiperidinium chloride,4-cyano-piperidine hcl,4-cyano-piperidine hydrochloride,4-cyanopiperidine hydrochloride salt,4-piperidinecarbonitrile hydrochloride PubChem CID: 22744637 IUPAC Name: piperidine-4-carbonitrile;hydrochloride SMILES: C1CNCCC1C#N.Cl