accessibility menu, dialog, popup

UserName

H3COCH3SH3C

CAS RN 2530-10-1

IUPAC Name: 1-(2,5-dimethylthiophen-3-yl)ethan-1-one
Synonyms: 1-(2,5-dimethylthiophen-3-yl)ethanone
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: PUSJAEJRDNPYKM-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)SC(C)=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)