CAS RN 25459-12-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-Nitro-5-desnitro Tinidazole, TRC
CAS: 25459-12-5 Molecular Formula: C8 H13 N3 O4 S Molecular Weight (g/mol): 247.27 Synonym: 1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole,1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-imidazole,Tinidazole Imp. B (EP),Tinidazole RC B (USP) IUPAC Name: 1-(2-ethylsulfonylethyl)-2-methyl-4-nitroimidazole SMILES: CCS(=O)(=O)CCn1cc(nc1C)[N+](=O)[O-]