CAS RN 2550-75-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alodane, TRC
CAS: 2550-75-6 Molecular Formula: C9 H6 Cl8 Molecular Weight (g/mol): 397.77 Synonym: 2-Norbornene, 1,2,3,4,7,7-hexachloro-5,6-bis(chloromethyl)- (6CI,7CI,8CI),1,2,3,4,7,7-Hexachloro-5,6-bis(chloromethyl)bicyclo[2.2.1]hept-2-ene,1,2,3,4,7,7-Hexachloro-5,6-bis(chloromethyl)-2-norbornene,1,2,3,4,7,7-Hexachloro-5,6-di(chloromethyl)[2.2.1]bicyclohept-2-ene,AC 12402,Alodan,Alodan (pesticide),Chlorbicyclen,Cyclodan,Hercules 12402,Hercules 426,NSC 41891 IUPAC Name: (1R,4S)-1,2,3,4,7,7-hexachloro-5,6-bis(chloromethyl)bicyclo[2.2.1]hept-2-ene SMILES: ClCC1C(CCl)[C@]2(Cl)C(=C(Cl)[C@@]1(Cl)C2(Cl)Cl)Cl