CAS RN 25514-31-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Bruceantin A, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bruceine A, MedChemExpress
MedChemExpress Bruceine A(NSC310616; Dihydrobrusatol) is a natural quassinoid compound extracted from the dried fruits of Brucea javanica (L.