CAS RN 2555002-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzo[d]imidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Methoxy-2-(4-methoxy-3,5-dimethylpyridin-2-yl)-1H-benzimidazole, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.