CAS RN 2566-32-7
IUPAC Name:
3-methyl-2-(methylamino)butanoic acid hydrochloride
Synonyms:
3-methyl-2-(methylamino)butanoic acid hydrochloride
Molecular Weight (g/mol):
167.63
Molecular Formula:
C6H14ClNO2
InChi Key:
LLAKGSVMNLYWAQ-UHFFFAOYNA-N
SMILES:
Cl.CNC(C(C)C)C(O)=O
1
–
3
of
3
results
1
N-Methyl-dl-valine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Methyl-dl-valine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Methyl-dl-valine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.