accessibility menu, dialog, popup

UserName

NH2SSNN

CAS RN 25660-71-3

IUPAC Name: 5-(benzylsulfanyl)-1,3,4-thiadiazol-2-amine
Synonyms: 5-(benzylsulfanyl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 223.31
Molecular Formula: C9H9N3S2
InChi Key: BHIGBGKIAJJBGD-UHFFFAOYSA-N
SMILES: NC1=NN=C(SCC2=CC=CC=C2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results