CAS RN 25777-71-3
IUPAC Name:
2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate; methyl 2-methylprop-2-enoate
Synonyms:
methyl methacrylate crosspolymer
Molecular Weight (g/mol):
298.34
Molecular Formula:
C15H22O6
InChi Key:
XBDAKYZJVVYINU-UHFFFAOYSA-N
SMILES:
COC(=O)C(C)=C.CC(=C)C(=O)OCCOC(=O)C(C)=C