CAS RN 258278-28-3
IUPAC Name:
1,3-bis[2,6-bis(propan-2-yl)phenyl]-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium-2-ide
Synonyms:
1,3-bis(2,6-diisopropylphenyl)-4,5-dihydro-1λ⁵-imidazol-1-ylium-2-ide
Molecular Weight (g/mol):
390.62
Molecular Formula:
C27H38N2
InChi Key:
XZDYFCGKKKSOEY-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=[C-]1)C1=C(C=CC=C1C(C)C)C(C)C
1
–
1
of
1
results
1
SIPr (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.