CAS RN 25876-54-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7Beta-Hydroxy-4-cholesten-3-one, TRC
CAS: 25876-54-4 Molecular Formula: C27 H44 O2 Molecular Weight (g/mol): 400.64 Synonym: Cholest-4-en-3-one, 7-hydroxy-, (7β)-,Cholest-4-en-3-one, 7β-hydroxy- (8CI),(7β)-7-Hydroxycholest-4-en-3-one,δ4-7β-Hydroxycholestenone,7β-Hydroxy-4-cholesten-3-one,7β-Hydroxycholest-4-en-3-one IUPAC Name: (7S,8S,9S,10R,13R,14S,17R)-7-hydroxy-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)CC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C