CAS RN 2588-06-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phorate Oxon Sulfone, TRC
CAS: 2588-06-9 Molecular Formula: C7 H17 O5 P S2 Molecular Weight (g/mol): 276.31 Synonym: Phosphorothioic acid, O,O-diethyl S-[(ethylsulfonyl)methyl] ester,O,O-Diethyl S-(ethylsulfonyl)methyl phosphorothioate,PO-Phorate sulfone,Phorate oxon sulfone,Phoratoxon sulfone,Phorate-oxon-sulfone IUPAC Name: 1-[ethoxy(ethylsulfonylmethylsulfanyl)phosphoryl]oxyethane SMILES: CCOP(=O)(OCC)SCS(=O)(=O)CC