missing translation for 'onlineSavingsMsg'
Learn More

Phorate Oxon Sulfone, TRC

Product Code. 30041011
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041011 10 mg 10mg
18002440 2.5 mg 2.5mg
18071230 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041011 Supplier Toronto Research Chemicals Supplier No. TRCP35351510MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2588-06-9
Molecular Formula C7 H17 O5 P S2
Molecular Weight (g/mol) 276.31
Synonym Phosphorothioic acid, O,O-diethyl S-[(ethylsulfonyl)methyl] ester, O,O-Diethyl S-(ethylsulfonyl)methyl phosphorothioate, PO-Phorate sulfone, Phorate oxon sulfone, Phoratoxon sulfone, Phorate-oxon-sulfone
IUPAC Name 1-[ethoxy(ethylsulfonylmethylsulfanyl)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)SCS(=O)(=O)CC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H17O5PS2/c1-4-11-13(8,12-5-2)14-7-15(9,10)6-3/h4-7H2,1-3H3
Formula Weight 276.0255
Chemical Name or Material Phorate Oxon Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.