CAS RN 25889-60-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methylfluoranthene, TRC
CAS: 25889-60-5 Molecular Formula: C17 H12 Molecular Weight (g/mol): 216.28 Synonym: 1-Methyl-fluoranthene,1-Methylfluoranthene IUPAC Name: 1-methylfluoranthene SMILES: Cc1ccc2cccc3c4ccccc4c1c23