CAS RN 2589-73-3
IUPAC Name:
1-(4-hydroxyphenyl)octan-1-one
Synonyms:
1-(4-hydroxyphenyl)octan-1-one
Molecular Weight (g/mol):
220.31
Molecular Formula:
C14H20O2
InChi Key:
GPDYSJOGSNWMDZ-UHFFFAOYSA-N
SMILES:
CCCCCCCC(=O)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
4'-Hydroxyoctanophenone, 99%
CAS: 2589-73-3 Molecular Formula: C14H20O2 Molecular Weight (g/mol): 220.31 MDL Number: MFCD00082694 InChI Key: GPDYSJOGSNWMDZ-UHFFFAOYSA-N Synonym: 4-octanoylphenol,1-4-hydroxyphenyl octan-1-one,4'-hydroxyoctanophenone,1-octanone, 1-4-hydroxyphenyl,octanophenone, 4'-hydroxy,unii-e2e77z6duk,e2e77z6duk,1-4-hydroxyphenyl-1-octanone PubChem CID: 75767 IUPAC Name: 1-(4-hydroxyphenyl)octan-1-one SMILES: CCCCCCCC(=O)C1=CC=C(O)C=C1