CAS RN 25913-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tinoridine hydrochloride, MedChemExpress
MedChemExpress Tinoridine hydrochloride is a nonsteroidal anti-inflammatory drug and also has potent radical scavenger and antiperoxidative activity.