accessibility menu, dialog, popup

UserName

OH(R)O(S)OH(R)HO(S)HO

CAS RN 25990-60-7

IUPAC Name: (2S,3R,4S,5R)-oxane-2,3,4,5-tetrol
Synonyms: α-D-xylopyranose α-D-xylose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: SRBFZHDQGSBBOR-LECHCGJUSA-N
SMILES: O[C@@H]1CO[C@H](O)[C@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results