CAS RN 260264-93-5
IUPAC Name:
N-[2,2-dimethyl-1-(methylcarbamoyl)propyl]-2-[1-(N-hydroxyformamido)ethyl]-5-phenylpentanamide
Synonyms:
N-[2,2-dimethyl-1-(methylcarbamoyl)propyl]-2-[1-(N-hydroxyformamido)ethyl]-5-phenylpentanamide
Molecular Weight (g/mol):
391.51
Molecular Formula:
C21H33N3O4
InChi Key:
GHVMTHKJUAOZJP-UHFFFAOYNA-N
SMILES:
CNC(=O)C(NC(=O)C(CCCC1=CC=CC=C1)C(C)N(O)C=O)C(C)(C)C
1
–
2
of
2
results
2
GI254023X, MedChemExpress
MedChemExpress GI254023X is a potent MMP9 and ADAM10 inhibitor with IC50s of 2.5 and 5.3 nM, respectively.