CAS RN 260435-42-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N,N-Dimethyl-2-[5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine N-Oxide (Rizatriptan N-Oxide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Rizatriptan N10-Oxide, TRC
CAS: 260435-42-5 Molecular Formula: C15 H19 N5 O Molecular Weight (g/mol): 285.34 Synonym: 1H-Indole-3-ethanamine, N,N-dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-, N-oxide,N,N-Dimethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indole-3-ethanamine N-oxide,Rizatriptan N-Oxide IUPAC Name: N,N-dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine oxide SMILES: C[N+](C)([O-])CCc1c[nH]c2ccc(Cn3cncn3)cc12