CAS RN 260980-89-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Topilutamide, MedChemExpress
MedChemExpress Topilutamide is a topical nonsteroidal antiandrogen (NSAA).