accessibility menu, dialog, popup

UserName

NLiLiLi

CAS RN 26134-62-3

IUPAC Name: trilithium(1+) azanide
Synonyms: lithium nitride
Molecular Weight (g/mol): 36.84
Molecular Formula: H2Li3N
InChi Key: BHZCMUVGYXEBMY-UHFFFAOYSA-N
SMILES: [Li+].[Li+].[Li+].[NH2-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results