CAS RN 261506-27-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fulvestrant 3-Beta-D-Glucuronide, TRC
CAS: 261506-27-8 Molecular Formula: C38H55F5O9S Molecular Weight (g/mol): 782.89 Synonym: (7alpha,17beta)-17-Hydroxy-7-[9-[(4,4,5,5,5-pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-trien-3-yl beta-D-Glucopyranosiduronic Acid; SMILES: C[C@]12CC[C@H]3[C@@H]([C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)Cc4cc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)ccc34)[C@@H]1CC[C@@H]2O