Learn More
Fulvestrant 3-Beta-D-Glucuronide, TRC
Product Code. 30180426
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180426 10 mg 10mg
30180179 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30180426 Supplier Toronto Research Chemicals Supplier No. TRCF86251010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 261506-27-8
Molecular Formula C38H55F5O9S
Molecular Weight (g/mol) 782.89
Synonym (7alpha,17beta)-17-Hydroxy-7-[9-[(4,4,5,5,5-pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-trien-3-yl beta-D-Glucopyranosiduronic Acid;
SMILES C[C@]12CC[C@H]3[C@@H]([C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)Cc4cc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)ccc34)[C@@H]1CC[C@@H]2O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C38H55F5O9S/c1-36-17-15-26-25-12-11-24(51-35-32(47)30(45)31(46)33(52-35)34(48)49)21-23(25)20-22(29(26)27(36)13-14-28(36)44)10-7-5-3-2-4-6-8-18-53(50)19-9-16-37(39,40)38(41,42)43/h11-12,21-22,26-33,35,44-47H,2-10,13-20H2,1H3,(H,48,49)/t22-,26-,27+,28+,29-,30+,31+,32-,33+,35-,36+,53?/m1/s1
Formula Weight 782.3487
Chemical Name or Material Fulvestrant 3-beta-D-Glucuronide