CAS RN 26158-00-9
IUPAC Name:
benzenesulfonic acid hydrate
Synonyms:
benzenesulfonic acid hydrate
Molecular Weight (g/mol):
176.19
Molecular Formula:
C6H8O4S
InChi Key:
MVIOINXPSFUJEN-UHFFFAOYSA-N
SMILES:
O.OS(=O)(=O)C1=CC=CC=C1
1
–
3
of
3
results
1
Benzenesulfonic Acid Monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzenesulfonic Acid Monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benzenesulfonic Acid Monohydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.